MilliporeSigma
  • Overexpression of SLC25A38 protein on acute lymphoblastic leukemia cells.
Overexpression of SLC25A38 protein on acute lymphoblastic leukemia cells.
Oncology letters (2014-04-26)
Huaying Chen, Quanyi Lu, Yunwu Zhang, Cuilin Zhang, Han Zhang, Huaxi Xu
ABSTRACT
SLC25A38 is a recently identified protein that belongs to the mitochondrial solute carrier family, SLC25. Previous studies have shown that it is a pro-apoptotic protein, which regulates intrinsic caspase-dependent apoptosis. In order to clarify the effect of SLC25A38 protein expression on acute lymphoblastic leukemia (ALL) cells, we detected the expression of SLC25A38 in various cell lines (RPMI 8226, U266, Molt-4 and Jurkat) by western blot analysis. The results indicate that SLC25A38 is highly expressed in the four cell lines. Among 55 leukemia patients (adult, n=32 and infant, n=23), a high expression of SLC25A38 protein was observed in seven infant (7/23, 30.4%) and 15 adult (15/32, 46.9%) ALL patients. Two adult ALL patients that were positive for SLC25A38 were analyzed and the level of SLC25A38 significantly reduced or disappeared following combined chemotherapy, however, reappeared upon ALL recurrence. The expression level was identified to be associated with the proportion of blast cells in the bone marrow. Additionally, SLC25A38 and Notch1 were co-expressed in the four cell lines and the ALL patient samples. The present results show that expression of SLC25A38 is a common feature of ALL cells and may be a novel biomarker for diagnosis, as well as a potential therapeutic target for ALL.
Sigma-Aldrich
Anti-SLC25A38 antibody produced in rabbit, affinity isolated antibody