MilliporeSigma
  • Development of a (32)P-postlabeling/HPLC method for detection of dehydroretronecine-derived DNA adducts in vivo and in vitro.
Development of a (32)P-postlabeling/HPLC method for detection of dehydroretronecine-derived DNA adducts in vivo and in vitro.
Chemical research in toxicology (2001-02-15)
Y Yang, J Yan, M Churchwell, R Beger, P Chan, D R Doerge, P P Fu, M W Chou
ABSTRACT
Pyrrolizidine alkaloids are naturally occurring genotoxic chemicals produced by a large number of plants. Metabolism of pyrrolizidine alkaloids in vivo and in vitro generates dehydroretronecine (DHR) as a common reactive metabolite. In this study, we report the development of a (32)P-postlabeling/HPLC method for detection of (i) two DHR-3'-dGMP and four DHR-3'-dAMP adducts and (ii) a set of eight DHR-derived DNA adducts in vitro and in vivo. The approach involves (1) synthesis of DHR-3'-dGMP, DHR-3'-dAMP, and DHR-3',5'-dG-bisphosphate standards and characterization of their structures by mass and (1)H NMR spectral analyses, (2) development of optimal conditions for enzymatic DNA digestion, adduct enrichment, and (32)P-postlabeling, and (3) development of optimal HPLC conditions. Using this methodology, we have detected eight DHR-derived DNA adducts, including the two epimeric DHR-3',5'-dG-bisphosphate adducts both in vitro and in vivo.