MilliporeSigma
  • Formation of cis-coniferin in cell-free extracts of Fagus grandifolia Ehrh bark.
Formation of cis-coniferin in cell-free extracts of Fagus grandifolia Ehrh bark.
Plant physiology (1990-01-01)
E Yamamoto, E J Inciong, L B Davin, N G Lewis
ABSTRACT
American beech (Fagus grandifolia Ehrh) bark exclusively accumulates cis-monolignols and their glucosidic conjugates; no evidence for the accumulation of trans-monolignols has been found. The glucosyltransferase from this source exhibits a very unusual substrate specificity for cis, and not trans, monolignols. This is further evidence that cis monolignols are involved in lignin formation in these plant tissues. Preliminary evidence for the existence of a novel trans-cis monolignol isomerase was obtained, in agreement with our contention that this isomerization is not photochemically mediated.
Sigma-Aldrich
Abietin, ≥90% (LC/MS-ELSD)