MilliporeSigma
  • Inhibitors of NF-kappaB signaling: design and synthesis of a biotinylated isopanepoxydone affinity reagent.
Inhibitors of NF-kappaB signaling: design and synthesis of a biotinylated isopanepoxydone affinity reagent.
Bioorganic & medicinal chemistry letters (2002-11-07)
J Brad Shotwell, Brian Koh, Hui Won Choi, John L Wood, Craig M Crews
ABSTRACT
A number of inhibitors of NF-kappaB signaling arising from our recent syntheses of isopanepoxydone and panepoxydone have been identified. Structure-activity data have been correlated to allow the design and synthesis of an affinity reagent for the isolation and identification of any relevant cellular target.