MilliporeSigma
  • [Studies on the chemical constituents of Cornus officinalis Sieb et Zucc].
[Studies on the chemical constituents of Cornus officinalis Sieb et Zucc].
Yao xue xue bao = Acta pharmaceutica Sinica (1992-01-01)
S P Zhao, Z Xue
ABSTRACT
A new bisiridoid glucoside named cornuside was isolated from the fruits of Cornus officinalis Sieb et Zucc. The structure of cornuside has been established by spectroscopic methods (IR, MS, 1HNMR, 13CNMR).