MilliporeSigma
  • Simultaneous determination of levodopa and 3-O-methyldopa in human plasma by liquid chromatography with electrochemical detection.
Simultaneous determination of levodopa and 3-O-methyldopa in human plasma by liquid chromatography with electrochemical detection.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2004-03-17)
Christoph Saxer, Miyuki Niina, Akinori Nakashima, Yusuke Nagae, Naoki Masuda
ABSTRACT
A simple and rapid assay is described for the simultaneous analysis of levodopa (l-DOPA) and 3-O-methyldopa (3-OMD) in human plasma samples, applying an ion-pair reversed-phase liquid chromatographic method with electrochemical detection, designed for clinical trials performed to study the effect of peripheral catechol-O-methyltransferase inhibitors on the metabolism of l-DOPA. After protein precipitation of 100 microl plasma sample aliquots with perchloric acid, the analytes are directly injected, separated within 10 min and simultaneously quantified down to 20 ng/ml by an electrochemical detector equipped with a dual-electrode system operating in redox mode eliminating effectively potential endogenous and exogenous interferences. The intra-assay precision for l-DOPA and 3-OMD was 1.34-6.54 and 3.90-5.50%, whereas the inter-assay precision was 2.09-7.69 and 4.16-9.90%, respectively. The recoveries were close to 90% for l-DOPA and almost 100% for 3-OMD. Satisfactory storage stability was achieved for up to 16 weeks at -70 degrees C by stabilizing plasma samples with antioxidants.
Product Description
USP
Levodopa Related Compound B, United States Pharmacopeia (USP) Reference Standard