MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • [Mollusca contagiosa in an infant with atopic eczema. A therapeutic challenge].
[Mollusca contagiosa in an infant with atopic eczema. A therapeutic challenge].
Der Hautarzt; Zeitschrift fur Dermatologie, Venerologie, und verwandte Gebiete (2005-01-07)
S M Erdmann, A Rübben, J Frank, P Poblete-Gutiérrez
ABSTRACT
Molluscum contagiosum is a viral disease frequently observed in children and adolescents with atopic dermatitis, in whom it poses a special therapeutic challenge. A 3-year-old girl presented with atopic eczema and disseminated molluscum contagiosum on her buttocks and upper legs. Topical application of 0.05% tretinoin resulted in a steady regression of the lesions. Other therapeutic options include topical anti-viral agents, cryosurgery, laser treatment, and curettage. Further, the therapeutic outcome is highly dependent on an adequate therapy of the concomitant atopic eczema.
Product Description
Prednicarbate, European Pharmacopoeia (EP) Reference Standard
Prednicarbate for system suitability A, European Pharmacopoeia (EP) Reference Standard