MilliporeSigma
  • A validated method for the determination of nicotine, cotinine, trans-3'-hydroxycotinine, and norcotinine in human plasma using solid-phase extraction and liquid chromatography-atmospheric pressure chemical ionization-mass spectrometry.
A validated method for the determination of nicotine, cotinine, trans-3'-hydroxycotinine, and norcotinine in human plasma using solid-phase extraction and liquid chromatography-atmospheric pressure chemical ionization-mass spectrometry.
Journal of mass spectrometry : JMS (2006-05-18)
Insook Kim, Marilyn A Huestis
ABSTRACT
A liquid chromatographic-mass spectrometric method for the simultaneous determination of nicotine, cotinine, trans-3'-hydroxycotinine, and norcotinine in human plasma was developed and validated. Analytes and deuterated internal standards were extracted from human plasma using solid-phase extraction and analyzed by liquid chromatography/atmospheric pressure chemical ionization-mass spectrometric detection with selected ion monitoring (SIM). Limits of detection and quantification were 1.0 and 2.5 ng/ml, respectively, for all analytes. Linearity ranged from 2.5 to 500 ng/ml of human plasma using a weighting factor of 1/x; correlation coefficients for the calibration curves were > 0.99. Intra- and inter-assay precision and accuracy were < 15.0%. Recoveries were 108.2-110.8% nicotine, 95.8-108.7% cotinine, 90.5-99.5% trans-3'-hydroxycotinine, and 99.5-109.5% norcotinine. The method was also partially validated in bovine serum, owing to the difficulty of obtaining nicotine-free human plasma for the preparation of calibrators and quality control (QC) samples. This method proved to be robust and accurate for the quantification of nicotine, cotinine, trans-3'-hydroxycotinine, and norcotinine in human plasma collected in clinical studies of acute nicotine effects on brain activity and on the development of neonates of maternal smokers.