MilliporeSigma
  • Potent cytotoxic rocaglamide derivatives from the fruits of Amoora cucullata.
Potent cytotoxic rocaglamide derivatives from the fruits of Amoora cucullata.
Chemical & pharmaceutical bulletin (2006-09-02)
Parinuch Chumkaew, Shigeru Kato, Kan Chantrapromma
ABSTRACT
Two new rocaglamide derivatives, 1-O-formylrocagloic acid (1) and 3'-hydroxy rocagloic acid (2), together with five known compounds, rocaglaol (3), rocagloic acid (4), 3'-hydroxymethylrocaglate (5), 1-O-formylmethyl rocaglate (6), and methylrocaglate (7), were isolated from the fruits of Amoora cucullata. Their structures were elucidated by spectroscopic methods. Compounds 1-3, 6, and 7 exhibited potent cytotoxicity against KB, BC, and NCI-H187 cell lines, whereas 4 and 5 showed selective cytotoxicity against NCI-H187 cell line.
Sigma-Aldrich
Rocaglamide, ≥96% (HPLC)