MilliporeSigma
[Chemical constituents from root of Psammosilene tunicoides].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2007-07-28)
Xiao-Xiao Liu, Lei Wang, Qiang Wang, Bin Qiu
ABSTRACT
To investigate the chemical constituents from the root of Psammosilene tunicoides. Column chromatographic methods were used to isolate the chemical constituents of this plant. ESI-MS, EI-MS and NMR methods were employed for their structural elucidation. Seven compounds were isolated and identified as goyaprosaponin (1), Soya-cerebroside (2), tectoridin (3), alpha-spinasterol (4), tetracosanoic acid (5), beta-sitosterol (6), daucosterol (7) respectively. Compounds 2-7 were obtained from genus Psammosilene for the first time.