MilliporeSigma
  • Aryne acyl-alkylation in the general and convergent synthesis of benzannulated macrolactone natural products: an enantioselective synthesis of (-)-curvularin.
Aryne acyl-alkylation in the general and convergent synthesis of benzannulated macrolactone natural products: an enantioselective synthesis of (-)-curvularin.
Organic letters (2010-03-04)
Pamela M Tadross, Scott C Virgil, Brian M Stoltz
ABSTRACT
A general approach for the synthesis of benzannulated macrolactone natural products utilizing an aryne acyl-alkylation reaction is described. Toward this end, the total syntheses of the natural products (-)-curvularin, curvulin, and (-)-diplodialide C are reported. Furthermore, the aryne insertion technology has enabled the rapid conversion of simple diplodialide natural products to curvularin, thereby connecting these two biosynthetically distinct classes of compounds via synthetic methods.
Sigma-Aldrich
Curvularin, from Penicillium citrinum, ≥98% (HPLC)