MilliporeSigma
  • Purification and structure determination of gelatinase and collagenase inhibitors from Viola patrinii fermentation extracts.
Purification and structure determination of gelatinase and collagenase inhibitors from Viola patrinii fermentation extracts.
Immunopharmacology and immunotoxicology (2010-03-04)
Kyoung-Sook Kim, Yeon-Joo Kwak, Kyung-Mi Kim, Hai Yang Yu, Byoung-Won Kang, Eunsook Chung, Young-Choon Lee, Jung-In Kim, Jai-Heon Lee
ABSTRACT
Investigation of collagenase and gelatinase inhibitory natural components afforded two isoflavonoids. Two isoflavonoids, tectorigenin-7-O-β-D-glucoside (1) and luteolin-7-O-β-D-glucuronopyranoside (2), were isolated from ethyl acetate fraction of Viola patrinii fermentation extracts (VPFE). Of these, compounds 1 and 2 exhibited collagenase inhibitory activity (IC(50)) at a concentration of less than 1.5 μM, and compound 2 showed gelatinases A and B inhibitory activity (IC(50)) at 0.3 μM and 0.8 μM, respectively.