MilliporeSigma
  • Polymorphism in molecular solids: an extraordinary system of red, orange, and yellow crystals.
Polymorphism in molecular solids: an extraordinary system of red, orange, and yellow crystals.
Accounts of chemical research (2010-06-22)
Lian Yu
ABSTRACT
Diamond and graphite are polymorphs of each other: they have the same composition but different structures and properties. Many other substances exhibit polymorphism: inorganic and organic, natural and manmade. Polymorphs are encountered in studies of crystallization, phase transition, materials synthesis, and biomineralization and in the manufacture of specialty chemicals. Polymorphs can provide valuable insights into crystal packing and structure-property relationships. 5-Methyl-2-[(2-nitrophenyl)amino]-3-thiophenecarbonitrile, known as ROY for its red, orange, and yellow crystals, has seven polymorphs with solved structures, the largest number in the Cambridge Structural Database. First synthesized by medicinal chemists, ROY has attracted attention from solid-state chemists because it demonstrates the remarkable diversity possible in organic solids. Many structures of ROY polymorphs and their thermodynamic properties are known, making ROY an important model system for testing computational models. Though not the most polymorphic substance on record, ROY is extraordinary in that many of its polymorphs can crystallize simultaneously from the same liquid and are kinetically stable under the same conditions. Studies of ROY polymorphs have revealed a new crystallization mechanism that invalidates the common view that nucleation defines the polymorph of crystallization. A slow-nucleating polymorph can still dominate the product if it grows rapidly and nucleates on another polymorph. Studies of ROY have also helped understand a new, surprisingly fast mode of crystal growth in organic liquids cooled to the glass transition temperature. This growth mode exists only for those polymorphs that have more isotropic, and perhaps more liquid-like, packing. The rich polymorphism of ROY results from a combination of favorable thermodynamics and kinetics. Not only must there be many polymorphs of comparable energies or free energies, many polymorphs must be kinetically stable and crystallize at comparable rates to be observed. This system demonstrates the unique insights that polymorphism provides into solid-state structures and properties, as well as the inadequacy of our current understanding of the phenomenon. Despite many studies of ROY, it is still impossible to predict the next molecule that is equally or more polymorphic. ROY is a lucky gift from medicinal chemists.
Product Description
USP
Olanzapine Related Compound A, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
Olanzapine impurity A, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.