MilliporeSigma
  • Dopexamine has no additional benefit in high-risk patients receiving goal-directed fluid therapy undergoing major abdominal surgery.
Dopexamine has no additional benefit in high-risk patients receiving goal-directed fluid therapy undergoing major abdominal surgery.
Anesthesia and analgesia (2010-11-05)
Simon J Davies, David Yates, R Jonathan T Wilson
ABSTRACT
Dopexamine has been shown to reduce both mortality and morbidity in major surgery when it is used as part of a protocol to increase oxygen delivery in the perioperative period. A European multicenter study has examined the use of dopexamine in patients undergoing major abdominal surgery, showing a trend toward improved survival and reduced complications in high-risk patients when receiving low-dose dopexamine (0.5 μg · kg(-1) · min(-1)). A reduced oxygen uptake at the anaerobic threshold (AT) has been shown to confer a significant risk of mortality in patients undergoing major abdominal surgery and allows objective identification of a high-risk operative group. In this study, we assessed the effects of low-dose dopexamine on morbidity after major abdominal surgery in patients who were at increased risk by virtue of a reduced AT. Patients undergoing elective major colorectal or urological surgery who had an AT of <11 mL · kg(-1) · min(-1) or an AT of 11 to 14 mL · kg(-1) · min(-1) with a history of ischemic heart disease were recruited. Before surgery, a radial arterial cannula was placed and attached to an Edwards Lifesciences FloTrac/Vigileo system for measuring cardiac output. Patients were given a 250-mL bolus of Voluven (6% hydroxyethyl starch 130/0.4 in 0.9% sodium chloride) until the stroke volume no longer increased by 10%, then received either dopexamine (0.5 μg · kg(-1) · min(-1)) or saline 0.9% for 24 hours. During surgery, fluid boluses of Voluven were given if the stroke volume variation was >10%. No crystalloid was given during surgery. A standardized postoperative fluid regime with Hartmann solution was prescribed at 1.5 mL · kg(-1) · h(-1) for 24 hours. The primary outcome measure was postoperative morbidity measured by the Postoperative Morbidity Survey. One hundred twenty-four patients were recruited over a 23-month period. The incidence of morbidity as measured by the Postoperative Morbidity Survey on day 5 was 55% in the control group versus 47% in the dopexamine group (P = 0.14). There was no significant reduction in morbidity on any measured postoperative day. Complication rates, mortality, and hospital length of stay were similar between the 2 groups; however, administration of dopexamine was associated with earlier return of tolerating an enteral diet. With the effective use of goal-directed fluid therapy in elective surgical patients, the routine use of dopexamine does not confer an additional clinical benefit.
Product Description
Dopexamine dihydrochloride, European Pharmacopoeia (EP) Reference Standard