MilliporeSigma
  • Interference of acute cigarette smoking with [¹³C]methacetin breath test.
Interference of acute cigarette smoking with [¹³C]methacetin breath test.
Isotopes in environmental and health studies (2011-02-03)
Anna Kasicka-Jonderko, Dorota Loska, Krzysztof Jonderko, Magdalena Kaminska, Barbara Błonska-Fajfrowska
ABSTRACT
It is essential to establish whether and how environmental factors affect the reliability of [(13)C]methacetin breath test ((13)C-MBT). In 12 healthy volunteers (smokers), a standard (13)C-MBT with 75 mg [(13)C]methacetin was performed twice in random order: on a control day without smoking and on another day with smoking two cigarettes antecedently. A considerable flattening of the curve of the momentary (13)C recovery within the expiratory air was observed when the (13)C-MBT was performed after smoking. The maximum of the momentary (13)C recovery, D(max), decreased from 37.20±2.58 to 25.39±2.29% dose/h (p=0.00052). Moreover, the time to reach D(max) was prolonged after cigarette smoking (26.5±3.1 vs. 16.5±1.9 min, p=0.0199). The curve of the cumulative (13)C recovery on the cigarette smoking day appeared to be shifted downwards, and statistically significant differences relative to the control situation were found between the 24th and 75th minute following [(13)C]methacetin administration. Smoking cigarettes immediately prior to the (13)C-MBT diminishes the ability of the liver to handle methacetin, and hence a possibility of such an interaction should be excluded in order to interpret the results of the test correctly.