MilliporeSigma
  • Formal synthesis of nitidine and NK109 via palladium-catalyzed domino direct arylation/N-arylation of aryl triflates.
Formal synthesis of nitidine and NK109 via palladium-catalyzed domino direct arylation/N-arylation of aryl triflates.
Organic letters (2011-02-26)
Mathieu Blanchot, David A Candito, Florent Larnaud, Mark Lautens
ABSTRACT
The use of aryl triflates as reaction partners in a palladium-catalyzed domino direct arylation/N-arylation provides a great advantage due to the availability of starting materials. Furthermore, it allows expedient access to biologically interesting benzo[c]phenanthridine alkaloids.