MilliporeSigma
  • Pharmacokinetics and pharmacodynamics of A77 1726 and leflunomide in domestic cats.
Pharmacokinetics and pharmacodynamics of A77 1726 and leflunomide in domestic cats.
Journal of veterinary pharmacology and therapeutics (2011-05-28)
M L Mehl, L Tell, A E Kyles, Y-J Chen, A Craigmill, C R Gregory
ABSTRACT
The pharmacokinetics and pharmacodynamics of A77 1726 and leflunomide after intravenous (i.v.) and oral (p.o.) administration were evaluated in adult cats. Three treatments were administered: a single i.v. dose of A77 1726 (4 mg/kg), a single oral dose of leflunomide (4 mg/kg), and multiple oral doses of leflunomide (2 mg/kg). Mean pharmacokinetic parameter values after a single i.v. dose of A77 1726 were distribution (A) and elimination (B) intercepts (15.2 μg/mL and 34.5 μg/mL, respectively), distribution and elimination half-lives (1.5 and 71.8 h, respectively), area under the curve (AUC(0 → ∞); 3723 μg*h/mL), mean residence time (MRT; 93 h), clearance (Cl(obs); 1.1 mL/kg/h), and volume of distribution at steady state (Vd(ss); 97 mL/kg). Mean pharmacokinetic parameter values after a single oral dose of leflunomide were absorption and elimination rate constants (0.3 1/h and 0.01 1/h, respectively), absorption and elimination half-lives (2.3 and 59.1 h, respectively), AUC(0 → ∞) (3966 μg*h/mL), and maximum observed plasma concentration (C(max); 38 μg/mL). The bioavailability after a single oral dose of leflunomide was 100%. The mean ± SD A77 1726 concentration that inhibited 50% lymphocytes (EC(50) ) was 16 ± 13.5 μg/mL. The mean ± SD maximum A77 1726 concentration (EC(max)) was 61.0 ± 23.9 μg/mL.
Product Description
USP
Leflunomide Related Compound B, United States Pharmacopeia (USP) Reference Standard