MilliporeSigma
  • Intraventricular hemorrhage in preterm infants: coagulation perspectives.
Intraventricular hemorrhage in preterm infants: coagulation perspectives.
Seminars in thrombosis and hemostasis (2011-12-22)
Amir A Kuperman, Gili Kenet, Emmanuel Papadakis, Benjamin Brenner
ABSTRACT
It has long been considered that a severe coagulation deficiency in premature newborns could be a major contributing factor in the occurrence of intraventricular hemorrhage (IVH). High-grade IVH has also been shown to coincide with severe derangement of coagulation in extremely low birth weight infants. This review focuses on the relevance of the physiologically developing immature hemostatic system to IVH, and the potential benefit of agents affecting hemostasis for IVH therapy or prevention in preterm infants. The findings of small, open-label interventional studies on the effect of ethamsylate, vitamin K, fresh frozen plasma, recombinant activated factor VII, and prothrombin complex concentrate on the premature coagulation system will be reviewed.
Product Description
Etamsylate, European Pharmacopoeia (EP) Reference Standard