MilliporeSigma
Diastereoselective synthesis of vicinal amino alcohols.
Organic & biomolecular chemistry (2012-04-27)
Oskari K Karjalainen, Ari M P Koskinen
ABSTRACT
The vicinal amino alcohol is a common motif in natural products and pharmaceuticals. Amino acids constitute a natural, inexpensive, and enantiopure choice of starting material for the synthesis of such functionalities. However, the matters concerning diastereoselectivity are not obvious. This Perspective takes a look in the field of diastereoselective synthesis of vicinal amino alcohols starting from amino acids using various methods.