- Immobilization of a thermostable cellobiose 2-epimerase from Rhodothermus marinus JCM9785 and continuous production of epilactose.
Immobilization of a thermostable cellobiose 2-epimerase from Rhodothermus marinus JCM9785 and continuous production of epilactose.
Bioscience, biotechnology, and biochemistry (2012-08-11)
Hiroki Sato, Wataru Saburi, Teruyo Ojima, Hidenori Taguchi, Haruhide Mori, Hirokazu Matsui
PMID22878201
ABSTRACT
Cellobiose 2-epimerase (CE) efficiently forms epilactose which has several beneficial biological functions. A thermostable CE from Rhodothermus marinus was immobilized on Duolite A568 and packed into a column. Lactose (100 g/L) was supplied to the reactor, kept at 50 °C at a space velocity of 8 h(-1). The epilactose concentration of the resulting eluate was 30 g/L, and this was maintained for 13 d.