MilliporeSigma
  • Multivariate spatial covariance analysis of 99mTc-exametazime SPECT images in dementia with Lewy bodies and Alzheimer's disease: utility in differential diagnosis.
Multivariate spatial covariance analysis of 99mTc-exametazime SPECT images in dementia with Lewy bodies and Alzheimer's disease: utility in differential diagnosis.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism (2013-01-31)
Sean J Colloby, John-Paul Taylor, Christopher M Davison, Jim J Lloyd, Michael J Firbank, Ian G McKeith, John T O'Brien
ABSTRACT
We examined (99m)Tc-exametazime brain blood flow single-photon emission computed tomography (SPECT) images using a spatial covariance analysis (SCA) approach to assess its diagnostic value in distinguishing dementia with Lewy bodies (DLB) from Alzheimer's disease (AD). Voxel SCA was simultaneously applied to a set of preprocessed images (AD, n=40; DLB, n=26), generating a series of eigenimages representing common intercorrelated voxels in AD and DLB. Linear regression derived a spatial covariance pattern (SCP) that discriminated DLB from AD. To investigate the diagnostic value of the model SCP, the SCP was validated by applying it to a second, independent, AD and DLB cohort (AD, n=34; DLB, n=29). Mean SCP expressions differed between AD and DLB (F(1,64)=36.2, P<0.001) with good diagnostic accuracy (receiver operating characteristic (ROC) curve area 0.87, sensitivity 81%, specificity 88%). Forward application of the model SCP to the independent cohort revealed similar differences between groups (F(1,61)=38.4, P<0.001), also with good diagnostic accuracy (ROC 0.86, sensitivity 80%, specificity 80%). Multivariate analysis of blood flow SPECT data appears to be robust and shows good diagnostic accuracy in two independent cohorts for distinguishing DLB from AD.
Product Description
Meso-rich exametazime, European Pharmacopoeia (EP) Reference Standard