MilliporeSigma
  • [Identification of metabolites of arbidol by ultra-high performance liquid chromatography tandem mass spectrometry].
[Identification of metabolites of arbidol by ultra-high performance liquid chromatography tandem mass spectrometry].
Yao xue xue bao = Acta pharmaceutica Sinica (2013-02-07)
Xiao Liu, Huan Li, Kai-Shun Bi, Xiao-Hui Chen, Hao Cai, Bao-Chang Cai
ABSTRACT
UPLC-MS-MS system was used for the identification of arbidol metabolites in the rat feces, urine and plasma samples. The system was so powerful a way with high ability of separation and analysis, based on both chromatography and mass properties. The isotope of Br was also a good indicator for metabolites finding. There were altogether 9 metabolites detected and identified, including 2 phase I biotransformation products and 7 phase II ones. It is concluded that arbidol mainly undergo metabolic reactions such as N-demethylation, S-oxidation, glucuronidation and sulfation in rats.
Sigma-Aldrich
Arbidol hydrochloride, ≥98% (HPLC)