MilliporeSigma
  • Protection of montelukast on OVA-induced eosinophilic gastroenteritis via modulating IL-5, eotaxin-1 and MBP expression.
Protection of montelukast on OVA-induced eosinophilic gastroenteritis via modulating IL-5, eotaxin-1 and MBP expression.
Immunopharmacology and immunotoxicology (2013-07-17)
Daihong Wan, Xiaohui Zhang, Xuemei Liu, Guimei Li
ABSTRACT
The aim of this study was to further explore the possible mechanisms of montelukast on oral mice ovalbumin-induced eosinophilic gastroenteritis in a mouse model. The results indicated that montelukast could prevent levels of eotaxin-1 and interleukin-5 in intestinal mucosa homogenate when compared with model group. Interestingly, the increase of major basic protein expression in jejunal tissue also was attenuated by treated with montelukast.
Product Description
Montelukast sodium, European Pharmacopoeia (EP) Reference Standard