MilliporeSigma
A case of Tsutsugamushi disease presenting with nephrotic syndrome.
The Korean journal of internal medicine (2013-12-07)
Ju Hyun Lee, Mi Jung Lee, Dong Ho Shin, Shin-Wook Kang, Kyu Hun Choi, Tae-Hyun Yoo
ABSTRACT
Nephrotic syndrome associated with Tsutsugamushi disease has not been previously reported. We are describing a case of Tsutsugamuchi disease presenting with nephrotic syndrome. A 72-year-old woman presented with fever and generalized edema. Laboratory studies revealed a leukocytosis, hypoalbuminemia, and hypercholesterolemia. Her urine protein excretion was 5.4 g/day. The anti-Tsutsugamushi antibody test was strongly positive (1:2,560). A renal biopsy was performed, and pathologic findings revealed membranous glomerulonephritis. The patient's clinical symptoms improved markedly after treatment with doxycycline.