MilliporeSigma
  • Metoclopramide does not increase gastric muscle contractility in newborn rats.
Metoclopramide does not increase gastric muscle contractility in newborn rats.
American journal of physiology. Gastrointestinal and liver physiology (2014-01-11)
Moshe Yair Kasirer, Christopher Welsh, Jingyi Pan, Yulia Shifrin, Jaques Belik
ABSTRACT
Feeding intolerance resulting from delayed gastric emptying is common in premature neonates. Metoclopramide (MCP), the most frequently used prokinetic drug in neonates, enhances gastric muscle contractility through inhibition of dopamine receptors. Although its therapeutic benefit is established in adults, limited data are available to support its clinical use in infants. Hypothesizing that developmentally dependent differences are present, we comparatively evaluated the effect of MCP on fundus muscle contractility in newborn, juvenile, and adult rats. The muscle strips were either contracted with electrical field stimulation (EFS) to induce cholinergic nerve-mediated acetylcholine release or carbachol, a cholinergic agonist acting directly on the muscarinic receptor. Although in adult rats MCP increased EFS-induced contraction by 294 ± 122% of control (P < 0.01), no significant effect was observed in newborn fundic muscle. MCP had no effect on the magnitude of the carbachol-induced and/or bethanechol-induced gastric muscle contraction at any age. In response to dopamine, an 80.7 ± 5.3% relaxation of adult fundic muscle was observed, compared with only a 8.4 ± 8.7% response in newborn tissue (P < 0.01). Dopamine D2 receptor expression was scant in neonates and significantly increased in adult gastric tissue (P < 0.01). In conclusion, the lack of MCP effect on the newborn fundic muscle contraction potential relates to developmental differences in dopamine D2 receptor expression. To the extent that these novel data can be extrapolated to neonates, the therapeutic value of MCP as a prokinetic agent early in life requires further evaluation.
Product Description
USP
Metoclopramide hydrochloride, United States Pharmacopeia (USP) Reference Standard