MilliporeSigma
Free radicals and their interpretations.
Acta physiologica Hungarica (1989-01-01)
T Jávor, T Past, L Nagy, G Mózsik, I Wittmann
ABSTRACT
Oxygen free radicals can be blamed for evoking gastric mucosal damage, because of the protective effect of some lipid soluble free radical scavengers (vitamin A related compounds, Vitamin E). Direct determination of free oxygen radicals related chemical entities in the gastric tissue during ulcerogenesis yielded controversial results. Aluminum antacid compound together with acid binding property exhibited cytoprotection too, elevating the tissue PGE2 level substantially. Magnesium containing antacid according to our model experiments on red blood cells damage by free radicals, is capable to bind free radicals as well as to counteract with the dangerous intracellular calcium accumulation. It has been concluded that aluminum-magnesium antacid has a cytoprotective effect via: 1. acid binding; 2. prostaglandin generation; 3. free radical scavenging; 4. calcium antagonist activity.
Product Description
Almagate, European Pharmacopoeia (EP) Reference Standard