MilliporeSigma
Clinical use of digitalis glycosides. An update.
Cardiology (1985-01-01)
J E Doherty
ABSTRACT
Digitalis glycosides continue to place high on the list of prescribed drugs. Digoxin is 8th on prescriptions written in the United States in 1980, digitoxin 16th, and digitalis leaf 23rd. There is little doubt that most physicians continue to believe these drugs are useful. The application of more definite indications, smaller doses, and the recognition of the role of pharmacokinetics and drug interactions make use of the glycosides more challenging than ever before in 1985.
Product Description
Deslanoside, European Pharmacopoeia (EP) Reference Standard