MilliporeSigma
  • Quantitative relationships between lipophilicity and mutagenic effects of N-substituted amides of 3-(5-nitro-2-furyl)-acrylic acid on Salmonella typhimurium.
Quantitative relationships between lipophilicity and mutagenic effects of N-substituted amides of 3-(5-nitro-2-furyl)-acrylic acid on Salmonella typhimurium.
Chemico-biological interactions (1985-10-01)
S Baláz, E Sturdík, I Dibus, L Ebringer, L Stibrányi, M Rosenberg
ABSTRACT
Mutagenic effects of 10 N-alkylamides of 3-(5-nitro-2-furyl)-acrylic acid were assessed in two strains of Salmonella typhimurium TA100 (rfa+ and rfa-). Experimental data were confronted with physiologically based compartment model comprising passive membrane transport, metabolical inactivation in cytoplasm and formation of a reactive intermediate which is responsible for receptor modification. The quantitative dependences observed between mutagenicity and lipophilicity indicate that, in both cases, the drug-receptor interaction takes place in a hydrophobic compartment localized in the cytoplasm. The mutagenic potency of some derivatives was influenced also by stericity.