MilliporeSigma
  • Treatment of uncomplicated gonorrhoea in women with talampicillin in a single oral 1.5 g dose.
Treatment of uncomplicated gonorrhoea in women with talampicillin in a single oral 1.5 g dose.
The British journal of venereal diseases (1982-06-01)
K C Mohanty, R B Roy
ABSTRACT
Four hundred and sixty women with uncomplicated gonorrhea were treated with a single oral dose of 1.5 g talampicillin. The success rate among those who attended for at least one follow-up examination after treatment was 99.14%. The antibiotic was well tolerated and no side effects were reported. Talampicillin given under supervision has proved to be the most effective and safest treatment in this area. A smaller dosage of talampicillin than ampicillin is required, the drug is better tolerated by the patient, and the time of nursing staff is saved.
Product Description
Talampicillin hydrochloride, European Pharmacopoeia (EP) Reference Standard