MilliporeSigma
  • Kinetics of the postinhibitory reactions of acetylcholinesterase poisoned by chiral isomalathion: a surprising nonreactivation induced by the RP stereoisomers.
Kinetics of the postinhibitory reactions of acetylcholinesterase poisoned by chiral isomalathion: a surprising nonreactivation induced by the RP stereoisomers.
Chemical research in toxicology (1993-01-01)
C E Berkman, S Ryu, D A Quinn, C M Thompson
ABSTRACT
Inhibitory (ki), spontaneous (k0), and oxime-mediated reactivation (k(oxime)) reaction kinetics for the four stereoisomers of isomalathion (SPRC,SPSC,RPRC, and RPSC) were determined against rat brain acetylcholinesterase (AChE). (SPRC)-Isomalathion was the most potent anticholinesterase agent and RPSC-isomalathion the least potent with racemic material approximately midway in activity. Following inhibition of rat brain AChE by (SPRC)- or (SPSC)-isomalathion, k0 and k(oxime) values were obtained that were comparable to (SP)-isoparathion methyl, indicating that the same mechanism of inhibition was shared, namely, formation of an O,S-dimethyl phosphorothiolated enzyme. Conversely, no appreciable reactivation occurred with or without oxime following inhibition of rat brain AChE by (RPSC)- or (RPRC)-isomalathion. This observation was not consistent with (RP)-isoparathion methyl, and a switch in inhibition mechanism to the loss of the thiomethyl moiety is suggested. The nonreactivation of rat brain AChE following inhibition by the (RP)-isomalathion stereoisomers is postulated to result from a mechanism involving either a beta-elimination of diethyl fumarate or displacement of the thiosuccinate moiety from the phosphate moiety.
Product Description
Malathion impurity A, European Pharmacopoeia (EP) Reference Standard