MilliporeSigma
  • New preparations of ethyl 3, 3-diethoxypropionate and ethoxycarbonylmalondialdehyde. Copper (I) catalyzed acetal formation from a conjugated triple bond.
New preparations of ethyl 3, 3-diethoxypropionate and ethoxycarbonylmalondialdehyde. Copper (I) catalyzed acetal formation from a conjugated triple bond.
The Journal of Organic Chemistry (1982)
Bertz SH, et al.