MilliporeSigma
Structure and inhibition of mouse leukotriene C4 synthase.
PloS one (2014-05-09)
Damian Niegowski, Thea Kleinschmidt, Shabbir Ahmad, Abdul Aziz Qureshi, Michaela Mårback, Agnes Rinaldo-Matthis, Jesper Z Haeggström
ABSTRACT
Leukotriene (LT) C4 synthase (LTC4S) is an integral membrane protein that catalyzes the conjugation reaction between the fatty acid LTA4 and GSH to form the pro-inflammatory LTC4, an important mediator of asthma. Mouse models of inflammatory disorders such as asthma are key to improve our understanding of pathogenesis and potential therapeutic targets. Here, we solved the crystal structure of mouse LTC4S in complex with GSH and a product analog, S-hexyl-GSH. Furthermore, we synthesized a nM inhibitor and compared its efficiency and binding mode against the purified mouse and human isoenzymes, along with the enzymes' steady-state kinetics. Although structural differences near the active site and along the C-terminal α-helix V suggest that the mouse and human LTC4S may function differently in vivo, our data indicate that mouse LTC4S will be a useful tool in future pharmacological research and drug development.