MilliporeSigma
  • Characterization of human phosphoserine aminotransferase involved in the phosphorylated pathway of L-serine biosynthesis.
Characterization of human phosphoserine aminotransferase involved in the phosphorylated pathway of L-serine biosynthesis.
The Biochemical journal (2003-03-14)
Joo Youn Baek, Do Youn Jun, Dennis Taub, Young Ho Kim
ABSTRACT
In the present study, we first report two forms of human phosphoserine aminotransferase (PSAT) cDNA (HsPSAT alpha and HsPSAT beta). HsPSAT alpha has a predicted open reading frame comprising 324 amino acids, encoding a 35.2 kDa protein (PSAT alpha), whereas HsPSAT beta consists of an open reading frame comprising 370 amino acids that encodes a 40 kDa protein (PSAT beta). PSAT alpha is identical with PSAT beta, except that it lacks 46 amino acids between Val(290) and Ser(337) of PSAT beta, which is encoded by the entire exon 8 (138 bp). Both PSAT alpha and PSAT beta can functionally rescue the deletion mutation of the Saccharomyces cerevisiae counterpart. Reverse transcriptase-PCR analysis revealed that the expression of PSAT beta mRNA was more dominant when compared with PSAT alpha mRNA in all human cell lines tested. PSAT beta was easily detected in proportion to the level of mRNA; however, PSAT alpha was detected only in K562 and HepG2 cells as a very faint band. The relative enzyme activity of glutathione S-transferase (GST)-PSAT beta expressed in Escherichia coli appeared to be 6.8 times higher than that of GST-PSAT alpha. PSAT mRNA was expressed at high levels (approx. 2.2 kb) in the brain, liver, kidney and pancreas, and very weakly expressed in the thymus, prostate, testis and colon. In U937 cells, the levels of PSAT mRNA and protein appeared to be up-regulated to support proliferation. Accumulation of PSAT mRNA reached a maximum in the S-phase of Jurkat T-cells. These results demonstrate that although two isoforms of human PSAT can be produced by alternative splicing, PSAT beta rather than PSAT alpha is the physiologically functional enzyme required for the phosphorylated pathway, and indicate that the human PSAT gene is regulated depending on tissue specificity as well as cellular proliferation status with a maximum level expression in the S-phase.