MilliporeSigma
  • A new approach to pyrrolophenanthridone alkaloids via allene intramolecular cycloaddition: Total synthesis of Hippadine.
A new approach to pyrrolophenanthridone alkaloids via allene intramolecular cycloaddition: Total synthesis of Hippadine.
Tetrahedron Letters (1987)
Hayakawa K, et al.