MilliporeSigma
  • Consolidated bioprocessing of highly concentrated Jerusalem artichoke tubers for simultaneous saccharification and ethanol fermentation.
Consolidated bioprocessing of highly concentrated Jerusalem artichoke tubers for simultaneous saccharification and ethanol fermentation.
Biotechnology and bioengineering (2013-04-10)
Lihao Guo, Jian Zhang, Fengxian Hu, Dewey Dy Ryu, Jie Bao
ABSTRACT
Consolidated bioprocessing (CBP) of Jerusalem artichoke tuber (Jat) for ethanol production is one of the most promising options for an alternate biofuel technology development. The technical barriers include the weak saccharolytic enzyme (inulinase) activity of the fermentation strain, and the well mixing of the high viscous fermentation slurry at the highly concentrated Jat loading. In this study, Saccharomyces cerevisiae DQ1 was found to produce relatively large amount of inulinase for hydrolysis of inulin in Jat, and the helical ribbon stirring bioreactor used provided well mixing performance under the high Jat loading. Even a highly concentrated Jat loading up to 35% (w/w) in the helical ribbon bioreactor for CBP was allowed. The results obtained from this study have demonstrated a feasibility of developing a CBP process technology in the helical ribbon bioreactor for ethanol production at a high yield 128.7 g/L and the theoretical yield 73.5%, respectively. This level of ethanol yield from Jat is relatively higher than others reported so far. The results of this study could provide a practical CBP process technology in the helical ribbon bioreactor for economically sustainable alternate biofuel production using highly concentrated inulin containing biomass feedstock such as Jat, at least 35%.
Sigma-Aldrich
Inulinase from Aspergillus niger, lyophilized, powder, brown-gray, ~25 U/mg