MilliporeSigma
  • In vitro kinetics of soybean lipoxygenase with combinatorial fatty substrates and its functional significance in off flavour development.
In vitro kinetics of soybean lipoxygenase with combinatorial fatty substrates and its functional significance in off flavour development.
Food chemistry (2013-11-02)
Somnath Mandal, Anil Dahuja, Abhijit Kar, I M Santha
ABSTRACT
Lipoxygenase (Lox) mediated oxidation of polyunsaturated fatty acids (PUFA) in mature soya seeds results in objectionable flavour. In the present study, Lox isozymes were purified to near homogeneity (107-fold). Lox-2 and 3 displayed remarkable kinetic preference (1.7 and 1.5-fold, respectively) for low PUFA ratios (LA/LeA) (PRs) among the selected PUFA combinations. Lox-1 displayed no specific preference. Pure Lox-1 displayed unbiased response towards substrates with marginal preference (1.2-fold) for linoleic acid at its optimum pH. Volatile compounds profiling showed a direct relationship between PRs and hexanal to trans-2-hexenal (1.47, 2.24 and 18.90 for 2, 7 and 15 PRs, respectively) ratio. The off-flavour determining parameters like TBA value, carbonyl value and lipid hydroperoxides (LHPODs) exhibited significant negative correlation (0.76, 0.74, 0.72; p<0.0001) in selected soya genotypes displaying varied PRs and significant positive correlation (0.89, 0.81. 0.89; p<0.0001) with ratio of PI (polyene index) to PRs - suggesting the plausible significance of PUFA ratios in biological lipid peroxidation.
Sigma-Aldrich
Lipoxidase from Glycine max (soybean), Type V, ammonium sulfate suspension, 500,000-1,000,000 units/mg protein
Sigma-Aldrich
Lipoxidase from Glycine max (soybean), Type I-B, lyophilized powder, ≥50,000 units/mg solid