MilliporeSigma
  • Transcription factors IIS and IIF enhance transcription efficiency by differentially modifying RNA polymerase pausing dynamics.
Transcription factors IIS and IIF enhance transcription efficiency by differentially modifying RNA polymerase pausing dynamics.
Proceedings of the National Academy of Sciences of the United States of America (2014-02-20)
Toyotaka Ishibashi, Manchuta Dangkulwanich, Yves Coello, Troy A Lionberger, Lucyna Lubkowska, Alfred S Ponticelli, Mikhail Kashlev, Carlos Bustamante
ABSTRACT
Transcription factors IIS (TFIIS) and IIF (TFIIF) are known to stimulate transcription elongation. Here, we use a single-molecule transcription elongation assay to study the effects of both factors. We find that these transcription factors enhance overall transcription elongation by reducing the lifetime of transcriptional pauses and that TFIIF also decreases the probability of pause entry. Furthermore, we observe that both factors enhance the processivity of RNA polymerase II through the nucleosomal barrier. The effects of TFIIS and TFIIF are quantitatively described using the linear Brownian ratchet kinetic model for transcription elongation and the backtracking model for transcriptional pauses, modified to account for the effects of the transcription factors. Our findings help elucidate the molecular mechanisms by which transcription factors modulate gene expression.