MilliporeSigma
  • Response surface optimization of enzymatic hydrolysis of Cistus ladanifer and Cytisus striatus for bio-ethanol production
Response surface optimization of enzymatic hydrolysis of Cistus ladanifer and Cytisus striatus for bio-ethanol production
Biochemical Engineering Journal (2009)
S. Ferreira et al.