MilliporeSigma
  • Effect of the consolute point of isobutyric acid+ water on the rate of an SN1 hydrolysis reaction.
Effect of the consolute point of isobutyric acid+ water on the rate of an SN1 hydrolysis reaction.
International Journal of Thermophysics (2001)
Kim YW and Baird JK.