MilliporeSigma
  • Methyl (E)-2-[(3-chloro-4-cyano-phenyl)-imino]-4-(4-chloro-phen-yl)-6-methyl-1,2,3,4-tetra-hydro-pyrimidine-5-carboxyl-ate.
Methyl (E)-2-[(3-chloro-4-cyano-phenyl)-imino]-4-(4-chloro-phen-yl)-6-methyl-1,2,3,4-tetra-hydro-pyrimidine-5-carboxyl-ate.
Acta crystallographica. Section E, Structure reports online (2012-11-06)
K N Venugopala, Susanta K Nayak, Bharti Odhav
ABSTRACT
In the title compound, C(20)H(16)Cl(2)N(4)O(2), the dihedral angles between the planes of the chloro-phenyl, chloro-cyano-phenyl-imine and ester groups and the plane of the six-membered tetra-hydro-pyrimidine ring are 86.9 (2), 72.6 (2) and 7.9 (2)°, respectively. The Cl atom substituent on the cyano-phenyl ring is disordered over two rotationally related sites [occupancy factors 0.887 (2):0.113 (2)], while the mol-ecular conformation is stabilized by the presence of an intra-molecular aromatic C-H⋯π inter-action. Both N-H groups participate in separate inter-molecular hydrogen-bonding associations with centrosymmetric cyclic motifs [graph sets R(2) (2)(8) and R(2) (2)(12)], resulting in ribbons parallel to [010]. Further weak C-H⋯O hydrogen bonds link these ribbons into a two-dimensional mol-ecular assembly.