MilliporeSigma
  • N-doped graphitic self-encapsulation for high performance silicon anodes in lithium-ion batteries.
N-doped graphitic self-encapsulation for high performance silicon anodes in lithium-ion batteries.
Energy & Environmental Science (2014)
JunaLee, Won, et al.