MilliporeSigma
  • Fabrication of thermosensitive polymer nanopatterns through chemical lithography and atom transfer radical polymerization.
Fabrication of thermosensitive polymer nanopatterns through chemical lithography and atom transfer radical polymerization.
Langmuir : the ACS journal of surfaces and colloids (2007-02-24)
Qiang He, Alexander Küller, Michael Grunze, Junbai Li
ABSTRACT
Micro- and nanopatterns of thermosensitive poly(N-isopropylacrylamide) brush on gold substrate were prepared by using chemical lithography combined with surface-initiated atom transfer radical polymerization. Self-assembled monolayers of 4'-nitro-1, 1'-biphenyl-4-thiol were structured by chemical lithography which produced cross-linked 4'-amino-1,1'-biphenyl-4-thiol monolayer within a nitro-terminated matrix. The terminal amino groups in monolayers were bounded with the surface initiator bromoisobutyryl bromide. After polymerization, the smallest size can reach to 70-nm line width and dots. The thermosensitivity of poly(N-isopropylacrylamide) brushes is demonstrated by contact angle measurement and fluid atomic force microscopy. This fabrication approach allows creating spatially defined polymer patterns and provides a simple and versatile method to construct complex micro- and nanopatterned polymer brushes with spatial and topographic control in a single step.