MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Comparative effectiveness of telaprevir-based triple therapy in patients with chronic hepatitis C.
Comparative effectiveness of telaprevir-based triple therapy in patients with chronic hepatitis C.
Mayo Clinic proceedings (2014-03-26)
Badr Al-Bawardy, W Ray Kim, John J Poterucha, John B Gross, Michael R Charlton, Joseph J Larson, Colin L Colby, Kathy Canterbury, Jennifer Warner, Terry M Therneau
ABSTRACT
To examine the effectiveness and tolerability of triple therapy with pegylated interferon (p-IFN), ribavirin (RBV), and telaprevir in patients with chronic hepatitis C receiving treatment in an academic practice setting and in a more clinically diverse population compared with patients receiving treatment in phase 3 trials. A prospective database of all patients with viral hepatitis undergoing antiviral therapy from January 1, 2006, to July 1, 2012, was queried to identify treatment-naive and -experienced patients with chronic hepatitis C receiving dual and triple therapies. On-treatment response categories included rapid virologic response, extended rapid virologic response, early virologic response, and sustained virologic response. These patients were compared with matched controls, namely, patients who underwent dual therapy with p-IFN and RBV. Matching was performed for age, cirrhosis status, and prior treatment. There were 55 patients who received triple therapy and met the eligibility criteria, consisting of treatment-naive (n=35) and -experienced patients (n=20: those with relapse, 9; those with nonresponse, 9; and those who terminated the treatment early, 2). Rapid virologic response was achieved in 41% of the patients, extended rapid virologic response in 41%, and early virologic response in 75%. Sustained virologic response was observed in 51% (18/35) of treatment-naive patients, 67% (6/9) of the patients with prior nonresponse, and 56% (5/9) of those with prior relapse. Corresponding results after dual therapy were 37% (23/62), 11% (2/18), and 27% (3/11), respectively. The mean decrease in the hemoglobin level at weeks 4, 8, and 24 of triple therapy was 2.8, 3.8, and 3.2 mg/dL compared with 2.4, 2.6, and 2.4 mg/dL with dual therapy (to convert mg/dL to mmol/L, multiply values by 0.0259). Telaprevir-based triple therapy in clinical practice is considerably more effective than dual therapy with p-IFN and RBV despite the significant degree of anemia that complicated therapy, requiring RBV dose reduction and erythropoietin support.
Product Description
Ribavirin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ribavirin, antiviral