MilliporeSigma
  • X-ray structure of human aquaporin 2 and its implications for nephrogenic diabetes insipidus and trafficking.
X-ray structure of human aquaporin 2 and its implications for nephrogenic diabetes insipidus and trafficking.
Proceedings of the National Academy of Sciences of the United States of America (2014-04-16)
Anna Frick, Urszula Kosinska Eriksson, Fabrizio de Mattia, Fredrik Oberg, Kristina Hedfalk, Richard Neutze, Willem J de Grip, Peter M T Deen, Susanna Törnroth-Horsefield
ABSTRACT
Human aquaporin 2 (AQP2) is a water channel found in the kidney collecting duct, where it plays a key role in concentrating urine. Water reabsorption is regulated by AQP2 trafficking between intracellular storage vesicles and the apical membrane. This process is tightly controlled by the pituitary hormone arginine vasopressin and defective trafficking results in nephrogenic diabetes insipidus (NDI). Here we present the X-ray structure of human AQP2 at 2.75 Å resolution. The C terminus of AQP2 displays multiple conformations with the C-terminal α-helix of one protomer interacting with the cytoplasmic surface of a symmetry-related AQP2 molecule, suggesting potential protein-protein interactions involved in cellular sorting of AQP2. Two Cd(2+)-ion binding sites are observed within the AQP2 tetramer, inducing a rearrangement of loop D, which facilitates this interaction. The locations of several NDI-causing mutations can be observed in the AQP2 structure, primarily situated within transmembrane domains and the majority of which cause misfolding and ER retention. These observations provide a framework for understanding why mutations in AQP2 cause NDI as well as structural insights into AQP2 interactions that may govern its trafficking.
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Cadmium, foil, not light tested, 25x25mm, thickness 0.01mm, 99.7+%
Cadmium, foil, 15mm disks, thickness 1.0mm, as rolled, 100%
Cadmium, wire reel, 5m, diameter 0.5mm, hard, 99.95+%
Cadmium, foil, 100x100mm, thickness 3.0mm, as rolled, 99.9%
Cadmium, foil, 10mm disks, thickness 0.125mm, as rolled, 99.95%
Cadmium, foil, 25x25mm, thickness 0.125mm, as rolled, 99.99+%
Cadmium, wire reel, 1m, diameter 1.2mm, as drawn, 99.99+%
Cadmium, foil, 4mm disks, thickness 0.01mm, 99.7+%
Cadmium, wire reel, 0.5m, diameter 1.0mm, hard, 99.999%
Cadmium, foil, 300x300mm, thickness 2.0mm, as rolled, 99.99%
Cadmium, foil, 15mm disks, thickness 0.25mm, as rolled, 99.99+%
Cadmium, wire reel, 0.2m, diameter 0.70mm, as drawn, 99.99+%
Cadmium, foil, 10mm disks, thickness 0.025mm, 99.7+%
Cadmium, wire reel, 10m, diameter 0.25mm, hard, 99.99+%
Cadmium, foil, 10mm disks, thickness 0.01mm, 99.7+%
Cadmium, foil, 30x100mm, thickness 0.5mm, as rolled, 99.999%
Cadmium, foil, not light tested, 50x50mm, thickness 0.005mm, 99.7+%
Cadmium, foil, not light tested, 25x25mm, thickness 0.005mm, 99.7+%
Cadmium, foil, 6mm disks, thickness 0.25mm, as rolled, 99.95%
Cadmium, foil, not light tested, 25x25mm, thickness 0.025mm, 99.7+%
Cadmium, foil, 8mm disks, thickness 0.005mm, 99.7+%
Cadmium, foil, 8mm disks, thickness 0.125mm, as rolled, 99.95%
Cadmium, foil, 6mm disks, thickness 1.0mm, as rolled, 100%
Cadmium, foil, not light tested, 25x25mm, thickness 0.015mm, 99.7+%
Cadmium, foil, 8mm disks, thickness 0.5mm, as rolled, 99.95%
Cadmium, foil, 6mm disks, thickness 0.05mm, as rolled, 99.99+%