MilliporeSigma
Post-traumatic stress symptoms and exacerbations in COPD patients.
COPD (2014-07-02)
Paulo Josè Zimermann Teixeira, Lucia Porto, Christian Haag Kristensen, Alvaro Huber Santos, Sergio Saldanha Menna-Barreto, Pedro AntÙnio Schmidt Do Prado-Lima
ABSTRACT
Post-Traumatic Stress Disorder (PTSD) is a common psychological consequence of exposure to traumatic stressful life events. During COPD exacerbations dyspnea can be considered a near-death experience that may induce post-traumatic stress symptoms. The aim of this study was to evaluate the relationship between COPD exacerbations and PTSD- related symptoms. Thirty-three in-patients with COPD exacerbations were screened for the following: PTSS (Screen for Posttraumatic Stress Symptoms), anxiety (Beck Anxiety Inventory) and depression (Beck Depression Inventory). Patients had a median age of 72 years and 72.7% were female. Mean FEV1 and FVC were 0.8 ± 0.3 (37.7 ± 14.9% of predicted) and 1.7 ± 0.6 (60 ± 18.8% of predicted), respectively with a mean exacerbation of 2.9 episodes over the past year. Post-traumatic stress symptoms related to PTSD were found in 11 (33.3%) patients (SPTSS mean score 4.13 ± 2.54); moderate to severe depression in 16 (48.5%) (BDI mean score 21.2 ± 12.1) and moderate to severe anxiety in 23 (69.7%) (BAI mean score 23.5 ± 12.4). In a linear regression model, exacerbations significantly predicted post-traumatic stress symptoms scores: SPTSS scores increased 0.9 points with each exacerbation (p = 0.001). Significant correlations were detected between PTSD-related symptoms and anxiety (rs = 0.57; p = 0.001) and PTSD symptoms and depression (rs = 0.62; p = 0.0001). In a multivariable analysis model, two or more exacerbation episodes led to a near twofold increase in the prevalence ratio of post-traumatic stress symptoms related to PTSD(PR1.71; p = 0.015) specially those requiring hospitalization (PR 1.13; p = 0.030) CONCLUSION: PTSD symptoms increase as the patient's exacerbations increase. Two or more exacerbation episodes lead to a near twofold increase in the prevalence ratio of post-traumatic symptomatology. Overall, these findings suggest that psychological domains should be addressed along with respiratory function and exacerbations in COPD patients.