MilliporeSigma
  • PLGA-PEG Encapsulated sitamaquine nanoparticles drug delivery system against Leishmania donovani
PLGA-PEG Encapsulated sitamaquine nanoparticles drug delivery system against Leishmania donovani
Journal of Scientific and Innovative Research (2014)
Kumara, R., Sahoo, G. C., Pandeya, K., Dasa, V. N. R., Yousuf, M., Ansaria, S. R., & Dasa, P.