MilliporeSigma
Immobilization of chloroperoxidase on aminopropyl-glass.
Applied and environmental microbiology (1990-11-01)
T A Kadima, M A Pickard
ABSTRACT
Chloroperoxidase (CPO) purified from Caldariomyces fumago CMI 89362 was covalently bound to aminopropyl-glass by using a modification of an established method. Acid-washed glass was derivatized by using aminopropyltriethoxysilane, and the enzyme was ionically bound at low ionic strength. Further treatment with glutaraldehyde covalently linked the enzyme to the glass beads in an active form. No elution of bound activity from glass beads could be detected with a variety of washings. The loading of enzyme protein to the glass beads was highest, 100 mg of CPO per g of glass, at high reaction ratios of CPO to glass, but the specific activity of the immobilized enzyme was highest, 36% of theoretical, at low enzyme-to-carrier ratios. No differences in the properties of the soluble and immobilized enzymes could be detected by a number of criteria: their pH-activity and pH-stability profiles were similar, as were their thermal stabilities. After five uses, the immobilized enzyme retained full activity between pH 6.0 and 6.7.