MilliporeSigma
  • Kinetics and mechanisms of the thermal decomposition of 2-methyl-1,3-dioxolane, 2,2-dimethyl-1,3-dioxolane, and cyclopentanone ethylene ketal in the gas phase. Combined experimental and DFT study.
Kinetics and mechanisms of the thermal decomposition of 2-methyl-1,3-dioxolane, 2,2-dimethyl-1,3-dioxolane, and cyclopentanone ethylene ketal in the gas phase. Combined experimental and DFT study.
The journal of physical chemistry. A (2012-08-14)
Felix Rosas, Jesus Lezama, José R Mora, Alexis Maldonado, Tania Cordova, Gabriel Chuchani
ABSTRACT
The kinetics of the gas-phase thermal decomposition of 2-methyl-1,3-dioxolane, 2,2-dimethyl-1,3-dioxolane, and cyclopentanone ethylene ketal were determined in a static system and the reaction vessel deactivated with allyl bromide. The decomposition reactions, in the presence of the free radical suppressor propene, are homogeneous, are unimolecular, and follow first-order law kinetics. The products of these reactions are acetaldehyde and the corresponding ketone. The working temperature range was 459-490 °C, and the pressure range was 46-113 Torr. The rate coefficients are given by the following Arrhenius equations: for 2-methyl-1,3-dioxolane, log k = (13.61 ± 0.12) - (242.1 ± 1.0)(2.303RT)(-1), r = 0.9997; for 2,2-dimethyl-1,3-dioxolane, log k = (14.16 ± 0.14) - (253.7 ± 2.0)(2.303RT)(-1), r = 0.9998; for cyclopentanone ethylene ketal, log k = (14.16 ± 0.14) - (253.7 ± 2.0)(2.303RT)(-1), r = 0.9998. Electronic structure calculations using DFT methods B3LYP and MPW1PW91 with 6-31G(d,p), and 6-31++G(d,p) basis sets suggest that the decomposition of these substrates takes place through a stepwise mechanism. The rate-determining step proceeds through a concerted nonsynchronous four-centered cyclic transition state, and the elongation of the C-OCH(3) bond in the direction C(α)(δ+)...OCH(3)(δ-) is predominant. The intermediate products of these decompositions are unstable, at the working temperatures, decomposing rapidly through a concerted cyclic six-centered cyclic transition state type of mechanism.
Sigma-Aldrich
2-Methyl-1,3-dioxolane, 97%