MilliporeSigma
  • Cholestanol metabolism, molecular pathology, and nutritional implications.
Cholestanol metabolism, molecular pathology, and nutritional implications.
Journal of medicinal food (2003-10-31)
Yousuke Seyama
ABSTRACT
Cholestanol, not cholesterol, is a minor component in the human body and in foods, but an increase in cholestanol concentration in serum induces a pathological condition named cerebrotendinous xanthomatosis (CTX). In our investigation of this disease for more than 25 years, a procedure for quantification of cholestanol by high-performance liquid chromatography and an assay method for sterol 27-hydroxylase were established, and several mutations of the CYP 27 gene in 10 CTX families were identified. We also established experimental animal models with symptoms of CTX by feeding a high cholestanol diet. Corneal dystrophy and gallstones were produced in mice, and an apoptosis of cerebellar neuronal cells was observed in rats. We propose the following underlying mechanism of CTX pathogenesis: When cholesterol in the plasma membrane is replaced by cholestanol to some extent, the membrane fluidity is reduced, and the calcium channel fails to open, inducing cell death. CTX patients are treated with oral administration of chenodeoxycholic acid, which reduces the cholestanol concentration in serum. Cholestanol has a toxic effect, and an imbalance of the cholesterol/cholestanol ratio in plasma membrane is suspected to cause the disturbance of calcium channel function of the membrane.
Sigma-Aldrich
5α-Cholestan-3β-ol, ≥95%