MilliporeSigma
Oxaprozin: a once-daily nonsteroidal anti-inflammatory drug.
Clinical pharmacy (1992-07-01)
L G Miller
ABSTRACT
The pharmacology, pharmacokinetics, clinical efficacy, adverse effects, and dosage of oxaprozin are reviewed. Oxaprozin, a nonsteroidal anti-inflammatory drug (NSAID) under consideration for approval by the Food and Drug Administration, is characterized as a propionic acid. By inhibiting cyclo-oxygenase, oxaprozin decreases the formation of prostaglandin (PG) precursors from arachidonic acid, resulting in decreased PG biosynthesis and reduced pain and inflammatory responses. Oxaprozin is well absorbed after oral administration, and peak plasma concentration is reached in three to six hours. Oxaprozin is primarily eliminated by urinary excretion of the unchanged drug. It has a long elimination half-life and persists in synovial fluid. In clinical studies, oxaprozin was equally or more effective than aspirin and as effective as naproxen in the treatment of rheumatoid arthritis. For treatment of osteoarthritis, oxaprozin was as effective as naproxen and more effective than aspirin or piroxicam. Studies have also shown oxaprozin to be effective therapy for juvenile rheumatoid arthritis and ankylosing spondylitis. Oxaprozin, like other NSAIDs, can cause gastrointestinal adverse effects. Other possible adverse effects include allergic reactions, analgesic nephropathy, hepatotoxicity, and increased bleeding times. For adults, the anticipated daily dosage is 600-1200 mg given as a single dose for rheumatoid arthritis, osteoarthritis, and analgesia. In children, oxaprozin 10-20 mg/kg/day has been used to treat juvenile rheumatoid arthritis. Oxaprozin is as effective as other NSAIDs and offers once-daily dosing; however, it does not offer any therapeutic advantage over other currently available NSAIDs.
Sigma-Aldrich
Oxaprozin, solid
Pricing and availability is not currently available.
USP
Oxaprozin, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.